siRNA Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017182.2 | NP_058878.1 | 9 | 1167 | ||
| XM_006253573.2 | XP_006253635.1 | 9 | 1205 | |||
| XM_006253574.2 | XP_006253636.1 | 9 | 1295 | |||
| XM_006253575.2 | XP_006253637.1 | 9 | 1196 | |||
| XM_006253576.2 | XP_006253638.1 | 9 | 1193 | |||
| GenBank mRNA | BC089093.1 | - | 9 | 1167 | 7126329 | |
| M99065.2 | - | 8 | 1008 | |||
| U79139.1 | - | 9 | 1112 | |||

H2afy
Genomic Map