siRNA Details
| Gene Symbol: AMBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001633.3 | NP_001624.1 | 6 | 857 | ||
| GenBank mRNA | AK290837.1 | - | 6 | 685 | ||
| AK303795.1 | - | 6 | 583 | |||
| AK310776.1 | - | 6 | 945 | |||
| AK310791.1 | - | 6 | 874 | |||
| AY544123.1 | - | 6 | 654 | |||
| BC041593.1 | - | 6 | 820 | 5433460 | ||
| X04225.1 | - | 6 | 666 | |||
| X04494.1 | - | 6 | 639 | |||

AMBP
Genomic Map