siRNA Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017184.1 | NP_058880.1 | 3 | 89 | ||
| XM_006249862.2 | XP_006249924.1 | 2 | 1596 | |||
| XM_006249864.1 | XP_006249926.1 | 2 | 175 | |||
| XM_006249865.2 | XP_006249927.1 | 5 | 218 | |||
| XM_006249866.2 | XP_006249928.1 | 5 | 149 | |||
| XM_006249868.2 | XP_006249930.1 | 3 | 184 | |||
| XM_006249869.2 | XP_006249931.1 | 3 | 108 | |||
| GenBank mRNA | FQ214889.1 | - | 5 | 196 | ||
| FQ215520.1 | - | 5 | 171 | |||
| FQ215574.1 | - | 5 | 169 | |||
| FQ215749.1 | - | 5 | 169 | |||
| FQ216094.1 | - | 5 | 169 | |||
| FQ216822.1 | - | 5 | 169 | |||
| FQ217411.1 | - | 5 | 169 | |||
| J04993.1 | - | 3 | 89 | |||

Tnni1
Genomic Map