siRNA Details
| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017185.1 | NP_058881.1 | 7 | 523 | ||
| GenBank mRNA | FQ216337.1 | - | 7 | 565 | ||
| FQ217026.1 | - | 8 | 565 | |||
| FQ217309.1 | - | 7 | 556 | |||
| FQ217454.1 | - | 8 | 573 | |||
| FQ217985.1 | - | 8 | 565 | |||
| FQ218058.1 | - | 7 | 597 | |||
| FQ223581.1 | - | 8 | 565 | |||
| FQ223772.1 | - | 8 | 571 | |||
| FQ224402.1 | - | 2 | 187 | |||
| FQ224418.1 | - | 2 | 96 | |||
| FQ224464.1 | - | 8 | 565 | |||
| FQ224489.1 | - | 8 | 565 | |||
| M73701.1 | - | 7 | 523 | |||

Tnni2
Genomic Map