siRNA Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019206.2 | NP_062079.2 | 1 | 152 | ||
| XM_008767561.1 | XP_008765783.1 | 1 | 156 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk10| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019206.2 | NP_062079.2 | 1 | 152 | ||
| XM_008767561.1 | XP_008765783.1 | 1 | 156 | |||