siRNA Details
| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017188.1 | NP_058884.1 | 2 | 282 | ||
| GenBank mRNA | BC062057.1 | - | 2 | 308 | 5598983 | |
| FQ214202.1 | - | 2 | 310 | |||
| U40999.1 | - | 2 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc119| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017188.1 | NP_058884.1 | 2 | 282 | ||
| GenBank mRNA | BC062057.1 | - | 2 | 308 | 5598983 | |
| FQ214202.1 | - | 2 | 310 | |||
| U40999.1 | - | 2 | 282 | |||