siRNA Details
| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017188.1 | NP_058884.1 | 5 | 695 | ||
| GenBank mRNA | BC062057.1 | - | 5 | 721 | 5598983 | |
| FQ214202.1 | - | 5 | 723 | |||
| U40999.1 | - | 5 | 695 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011676, AK131955, AF030169, BC001990.

Unc119
Genomic Map