siRNA Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017190.4 | NP_058886.1 | 12 | 1999 | ||
| XM_006228824.2 | XP_006228886.1 | 11 | 2029 | |||
| XM_006228825.2 | XP_006228887.1 | 11 | 2546 | |||
| XM_006228827.2 | XP_006228889.1 | 13 | 2126 | |||
| XM_008759137.1 | XP_008757359.1 | 11 | 2553 | |||
| XM_008759138.1 | XP_008757360.1 | 11 | 2022 | |||
| XM_008759139.1 | XP_008757361.1 | 12 | 2597 | |||
| GenBank mRNA | FQ212027.1 | - | 13 | 2060 | ||
| M11721.1 | - | 6 | 915 | |||
| M14871.1 | - | 11 | 1969 | |||
| M16800.1 | - | 12 | 1999 | |||
| M22357.1 | - | 12 | 1992 | |||
| M36702.1 | - | 6 | 915 | |||
| X05301.1 | - | 11 | 1962 | |||
| X06554.1 | - | 5 | 629 | |||

Mag
Genomic Map