siRNA Details
| Gene Symbol: Ppt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022502.2 | NP_071947.1 | 7, 8 | 728 | ||
| GenBank mRNA | BC126089.1 | - | 7, 8 | 728 | 8361877 | |
| FQ213474.1 | - | 7, 8 | 740 | |||
| FQ213911.1 | - | 740 | ||||
| L34262.1 | - | 726 | ||||

Ppt1
Genomic Map