siRNA Details
| Gene Symbol: Ppt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022502.2 | NP_071947.1 | 9 | 874 | ||
| GenBank mRNA | BC126089.1 | - | 9 | 874 | 8361877 | |
| FQ213474.1 | - | 9 | 886 | |||
| FQ213911.1 | - | 886 | ||||
| L34262.1 | - | 872 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppt1| Gene Symbol: Ppt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022502.2 | NP_071947.1 | 9 | 874 | ||
| GenBank mRNA | BC126089.1 | - | 9 | 874 | 8361877 | |
| FQ213474.1 | - | 9 | 886 | |||
| FQ213911.1 | - | 886 | ||||
| L34262.1 | - | 872 | ||||