siRNA Details
Gene Symbol: Adra1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017191.2 | NP_058887.2 | 1 | 632 | ||
XM_006252107.2 | XP_006252169.1 | 2 | 1617 | |||
XM_006252108.2 | XP_006252170.1 | 2 | 1615 | |||
XM_006252109.2 | XP_006252171.1 | 2 | 1616 | |||
XM_006252110.2 | XP_006252172.1 | 2 | 1611 | |||
XM_008770744.1 | XP_008768966.1 | 2 | 1617 | |||
GenBank mRNA | S67375.1 | - | 1 | 29 | ||
U07126.1 | - | 1 | 632 | |||
U13368.1 | - | 1 | 652 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
