siRNA Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133287.1 | NP_579821.1 | 4 | 332 | ||
| GenBank mRNA | AB210108.1 | - | 4 | 332 | ||
| BC166596.1 | - | 631 | 7934724 | |||
| DQ336702.1 | - | 5 | 366 | |||
| DQ336703.1 | - | 4 | 333 | |||
| DQ336704.1 | - | 4 | 333 | |||
| DQ336705.1 | - | 5 | 366 | |||

Numb
Genomic Map