siRNA Details
| Gene Symbol: Chrne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017194.1 | NP_058890.1 | 5 | 529 | ||
| XM_006246678.2 | XP_006246740.1 | 6 | 647 | |||
| XM_008767807.1 | XP_008766029.1 | 6 | 630 | |||
| XM_008767808.1 | XP_008766030.1 | 7 | 1296 | |||
| GenBank mRNA | X13252.1 | - | 5 | 529 | ||
| X74836.1 | - | 5 | 529 | |||

Chrne
Genomic Map