siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 3 | 613 | ||
| GenBank mRNA | D45247.1 | - | 3 | 611 | ||
| FQ214283.1 | - | 2 | 147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb5| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105727.2 | NP_001099197.2 | 3 | 613 | ||
| GenBank mRNA | D45247.1 | - | 3 | 611 | ||
| FQ214283.1 | - | 2 | 147 | |||