siRNA Details
| Gene Symbol: Celf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083586.2 | NP_001077055.2 | 2, 3 | 485 | ||
| NM_017197.3 | NP_058893.2 | 2, 3 | 485 | |||
| XM_008771861.1 | XP_008770083.1 | 2, 3 | 364 | |||
| XM_008771862.1 | XP_008770084.1 | 1, 2 | 922 | |||
| XM_008771863.1 | XP_008770085.1 | 1, 2 | 921 | |||
| XM_008771864.1 | XP_008770086.1 | 1, 2 | 921 | |||
| GenBank mRNA | AF090695.1 | - | 2, 3 | 319 | ||
| AJ010351.1 | - | 2, 3 | 410 | |||
| AJ010386.1 | - | 2, 3 | 533 | |||
| BC129096.1 | - | 2, 3 | 311 | 8362847 | ||

Celf2
Genomic Map