siRNA Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017198.1 | NP_058894.1 | 13 | 1691 | ||
| XM_006229752.1 | XP_006229814.1 | 13 | 1542 | |||
| XM_006229753.1 | XP_006229815.1 | 13 | 1539 | |||
| XM_006229754.2 | XP_006229816.1 | 13 | 1531 | |||
| GenBank mRNA | U23443.1 | - | 13 | 1497 | ||
| U49953.1 | - | 13 | 1691 | |||

Pak1
Genomic Map