siRNA Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017198.1 | NP_058894.1 | 12, 13 | 1595 | ||
| XM_006229752.1 | XP_006229814.1 | 12, 13 | 1446 | |||
| XM_006229753.1 | XP_006229815.1 | 12, 13 | 1443 | |||
| XM_006229754.2 | XP_006229816.1 | 12, 13 | 1435 | |||
| GenBank mRNA | U23443.1 | - | 12, 13 | 1401 | ||
| U49953.1 | - | 12, 13 | 1595 | |||

Pak1
Genomic Map