siRNA Details
| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019210.1 | NP_062083.1 | 5, 6 | 525 | ||
| XM_006257333.2 | XP_006257395.1 | 7, 8 | 1798 | |||
| XM_006257336.2 | XP_006257398.1 | 6, 7 | 1687 | |||
| XM_006257338.2 | XP_006257400.1 | 6, 7 | 2090 | |||
| XM_006257339.2 | XP_006257401.1 | 6, 7 | 1752 | |||
| XM_008773404.1 | XP_008771626.1 | 8, 9 | 1863 | |||
| GenBank mRNA | U33314.1 | - | 5, 6 | 525 | ||

Pak3
Genomic Map