siRNA Details
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019211.2 | NP_062084.2 | 9 | 1279 | ||
GenBank mRNA | AF060819.1 | - | 9 | 1141 | ||
AF081196.1 | - | 9 | 1279 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019211.2 | NP_062084.2 | 9 | 1279 | ||
GenBank mRNA | AF060819.1 | - | 9 | 1141 | ||
AF081196.1 | - | 9 | 1279 |