siRNA Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017199.1 | NP_058895.1 | 2, 3 | 336 | ||
| XM_006229571.2 | XP_006229633.1 | 2, 3 | 392 | |||
| GenBank mRNA | BC058482.1 | - | 2, 3 | 249 | 6921620 | |
| Z19087.1 | - | 2, 3 | 336 | |||

Ssr4
Genomic Map