siRNA Details
| Gene Symbol: Jtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019213.2 | NP_062086.1 | 3, 4 | 480 | ||
| GenBank mRNA | AB016489.1 | - | 3, 4 | 534 | ||
| BC062090.1 | - | 3, 4 | 480 | 5621612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jtb| Gene Symbol: Jtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019213.2 | NP_062086.1 | 3, 4 | 480 | ||
| GenBank mRNA | AB016489.1 | - | 3, 4 | 534 | ||
| BC062090.1 | - | 3, 4 | 480 | 5621612 | ||