siRNA Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019214.1 | NP_062087.1 | 7 | 986 | ||
| XM_008764576.1 | XP_008762798.1 | 8 | 2310 | |||
| XM_008764577.1 | XP_008762799.1 | 6 | 1077 | |||
| GenBank mRNA | AF167412.1 | - | 7 | 986 | ||

Slc26a4
Genomic Map