siRNA Details
| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031576.1 | NP_113764.1 | 1404 | |||
| GenBank mRNA | M10068.1 | - | 11 | 1368 | ||
| M12516.1 | - | 1404 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Por| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031576.1 | NP_113764.1 | 1404 | |||
| GenBank mRNA | M10068.1 | - | 11 | 1368 | ||
| M12516.1 | - | 1404 | ||||