siRNA Details
| Gene Symbol: Ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017201.1 | NP_058897.1 | 8 | 934 | ||
| GenBank mRNA | M15185.1 | - | 8 | 934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahcy| Gene Symbol: Ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017201.1 | NP_058897.1 | 8 | 934 | ||
| GenBank mRNA | M15185.1 | - | 8 | 934 | ||