siRNA Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017202.1 | NP_058898.1 | 3 | 313 | ||
| GenBank mRNA | BC084719.1 | - | 2 | 229 | 7317602 | |
| FQ211185.1 | - | 3 | 266 | |||
| FQ214265.1 | - | 3 | 322 | |||
| FQ217878.1 | - | 3 | 281 | |||
| FQ223981.1 | - | 3 | 275 | |||
| FQ227128.1 | - | 3 | 311 | |||
| FQ228805.1 | - | 3 | 263 | |||
| X14209.1 | - | 3 | 295 | |||
| X15029.1 | - | 2 | 240 | |||

Cox4i1
Genomic Map