siRNA Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017202.1 | NP_058898.1 | 2, 3 | 171 | ||
| GenBank mRNA | BC084719.1 | - | 1, 2 | 87 | 7317602 | |
| FQ211185.1 | - | 2, 3 | 124 | |||
| FQ214265.1 | - | 2, 3 | 180 | |||
| FQ217878.1 | - | 2, 3 | 139 | |||
| FQ223981.1 | - | 2, 3 | 133 | |||
| FQ227128.1 | - | 2, 3 | 169 | |||
| FQ228805.1 | - | 2, 3 | 121 | |||
| X14209.1 | - | 2, 3 | 153 | |||
| X15029.1 | - | 1, 2 | 98 | |||

Cox4i1
Genomic Map