siRNA Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017202.1 | NP_058898.1 | 4 | 344 | ||
| GenBank mRNA | BC084719.1 | - | 3 | 260 | 7317602 | |
| FQ211185.1 | - | 4 | 297 | |||
| FQ214265.1 | - | 4 | 353 | |||
| FQ217878.1 | - | 4 | 312 | |||
| FQ223981.1 | - | 4 | 306 | |||
| FQ227128.1 | - | 4 | 342 | |||
| FQ228805.1 | - | 4 | 294 | |||
| X14209.1 | - | 4 | 326 | |||
| X15029.1 | - | 3 | 271 | |||

Cox4i1
Genomic Map