siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053307.2 | NP_445759.1 | 2 | 182 | ||
| XM_008770745.1 | XP_008768967.1 | 2 | 553 | |||
| GenBank mRNA | AY005464.1 | - | 2 | 172 | ||
| BC087009.1 | - | 2 | 182 | 7107102 | ||
| DQ989019.1 | - | 3 | 214 | |||
| DQ989020.1 | - | 3 | 217 | |||

Msra
Genomic Map