siRNA Details
| Gene Symbol: Gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017203.1 | NP_058899.1 | 2 | 618 | ||
| GenBank mRNA | AF092733.1 | - | 2 | 423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf11| Gene Symbol: Gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017203.1 | NP_058899.1 | 2 | 618 | ||
| GenBank mRNA | AF092733.1 | - | 2 | 423 | ||