siRNA Details
| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017204.1 | NP_058900.1 | 5 | 3155 | ||
| XM_006229755.2 | XP_006229817.1 | 4 | 3485 | |||
| GenBank mRNA | X93495.1 | - | 5 | 3155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map6| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017204.1 | NP_058900.1 | 5 | 3155 | ||
| XM_006229755.2 | XP_006229817.1 | 4 | 3485 | |||
| GenBank mRNA | X93495.1 | - | 5 | 3155 | ||