siRNA Details
| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019219.2 | NP_062092.2 | 5 | 405 | ||
| GenBank mRNA | AF025819.2 | - | 5 | 405 | ||
| BC101915.1 | - | 5 | 449 | 7935765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp9| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019219.2 | NP_062092.2 | 5 | 405 | ||
| GenBank mRNA | AF025819.2 | - | 5 | 405 | ||
| BC101915.1 | - | 5 | 449 | 7935765 | ||