siRNA Details
| Gene Symbol: Vgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030997.1 | NP_112259.1 | 3 | 906 | ||
| XM_006249137.2 | XP_006249199.1 | 3 | 927 | |||
| XM_006249138.2 | XP_006249200.1 | 2 | 776 | |||
| XM_008769134.1 | XP_008767356.1 | 2 | 979 | |||
| GenBank mRNA | M60525.1 | - | 3 | 826 | ||
| M74223.1 | - | 2 | 772 | |||
| Gene Symbol: LOC100909521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257726.2 | XP_006257788.1 | 979 | |||

Vgf
Genomic Map