siRNA Details
| Gene Symbol: Avpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017205.3 | NP_058901.3 | 2, 3 | 1580 | ||
| GenBank mRNA | D45400.1 | - | 1, 2 | 1109 | ||
| U27322.1 | - | 1, 2 | 1473 | |||
| Gene Symbol: LOC100909648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289800.1 | NP_001276729.1 | 1, 2 | 1048 | ||
More Information
Set Membership: |
|

Avpr1b
Genomic Map