siRNA Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109986.1 | NP_001103456.1 | 4 | 323 | ||
| NM_024134.2 | NP_077048.2 | 3 | 275 | |||
| XM_006241444.2 | XP_006241506.1 | 5 | 1501 | |||
| XM_006241445.2 | XP_006241507.1 | 4 | 1453 | |||
| GenBank mRNA | AW916370.1 | - | 4 | 323 | ||
| BC100664.1 | - | 4 | 286 | 7932840 | ||
| CF977695.1 | - | 3 | 278 | |||
| U30186.1 | - | 3 | 267 | |||
| U36994.1 | - | 3 | 245 | |||

Ddit3
Genomic Map