siRNA Details
| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017319.1 | NP_059015.1 | 5 | 663 | ||
| GenBank mRNA | BC062393.1 | - | 5 | 700 | 5621329 | |
| D63378.1 | - | 5 | 663 | |||
| FQ211627.1 | - | 762 | ||||
| X12355.1 | - | 5 | 659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdia3| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017319.1 | NP_059015.1 | 5 | 663 | ||
| GenBank mRNA | BC062393.1 | - | 5 | 700 | 5621329 | |
| D63378.1 | - | 5 | 663 | |||
| FQ211627.1 | - | 762 | ||||
| X12355.1 | - | 5 | 659 | |||