siRNA Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019222.1 | NP_062095.1 | 10 | 1310 | ||
| XM_006230773.2 | XP_006230835.1 | 10 | 1429 | |||
| XM_006230774.2 | XP_006230836.1 | 10 | 1394 | |||
| GenBank mRNA | AJ006064.1 | - | 10 | 1310 | ||
| BC061558.1 | - | 9 | 1908 | 6919414 | ||
| FQ213235.1 | - | 6 | 683 | |||

Coro1b
Genomic Map