siRNA Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019222.1 | NP_062095.1 | 8 | 906 | ||
| XM_006230773.2 | XP_006230835.1 | 8 | 1025 | |||
| XM_006230774.2 | XP_006230836.1 | 8 | 990 | |||
| GenBank mRNA | AJ006064.1 | - | 8 | 906 | ||
| BC061558.1 | - | 7 | 1504 | 6919414 | ||
| FQ213235.1 | - | 4 | 279 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011778, BC132617, AK205268, AK008947, BC029326, and others.

Coro1b
Genomic Map