siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145005.1 | NP_001138477.1 | 2, 3 | 251 | ||
| NM_017213.2 | NP_058909.2 | 3, 4 | 446 | |||
| GenBank mRNA | AF053971.1 | - | 2, 3 | 547 | ||
| BC078857.1 | - | 3, 4 | 446 | 7112637 | ||
| CK470868.1 | - | 2, 3 | 251 | |||
| U62821.1 | - | 1, 2 | 278 | |||
| X95272.1 | - | 2, 3 | 217 | |||

Odf2
Genomic Map