siRNA Details
| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019224.2 | NP_062097.1 | 15 | 1210 | ||
| GenBank mRNA | AB019145.1 | - | 15 | 1210 | ||
| AF038006.1 | - | 6 | 536 | |||
| AF071475.1 | - | 15 | 1206 | |||
| U32433.1 | - | 2 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs9| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019224.2 | NP_062097.1 | 15 | 1210 | ||
| GenBank mRNA | AB019145.1 | - | 15 | 1210 | ||
| AF038006.1 | - | 6 | 536 | |||
| AF071475.1 | - | 15 | 1206 | |||
| U32433.1 | - | 2 | 100 | |||