siRNA Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035233.1 | NP_001030310.1 | 483 | |||
| NM_017215.2 | NP_058911.2 | 483 | ||||
| GenBank mRNA | AF297648.1 | - | 395 | |||
| AF465909.1 | - | 376 | ||||
| AY044832.1 | - | 372 | ||||
| U15098.1 | - | 432 | ||||
| X67857.1 | - | 470 | ||||

Slc1a2
Genomic Map