siRNA Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035233.1 | NP_001030310.1 | 938 | |||
| NM_017215.2 | NP_058911.2 | 938 | ||||
| GenBank mRNA | AF297648.1 | - | 850 | |||
| AF451299.1 | - | 310 | ||||
| AF465909.1 | - | 831 | ||||
| AY044832.1 | - | 827 | ||||
| AY069978.1 | - | 129 | ||||
| JN132400.1 | - | 1013 | ||||
| U15098.1 | - | 887 | ||||
| X67857.1 | - | 926 | ||||

Slc1a2
Genomic Map