siRNA Details
| Gene Symbol: SMARCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098426.1 | NP_001091896.1 | 9, 10 | 1500 | ||
| XM_005257604.2 | XP_005257661.2 | 9, 10 | 1047 | |||
| XM_017024967.1 | XP_016880456.1 | 9, 10 | 2239 | |||
| GenBank mRNA | AK025917.1 | - | 1, 2 | 64 | ||
| AK300939.1 | - | 9, 10 | 1284 | |||
| AK303951.1 | - | 9, 10 | 1086 | |||
| AK307944.1 | - | 9, 10 | 1053 | |||
| BC136322.1 | - | 10, 11 | 1369 | 9020309 | ||
| BC142963.1 | - | 9, 10 | 1449 | 8860380 | ||
| U66618.1 | - | 9, 10 | 1443 | |||

SMARCD2
Genomic Map