siRNA Details
| Gene Symbol: Ptbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271057.1 | NP_001257986.1 | 6 | 704 | ||
| NM_022516.5 | NP_071961.2 | 6 | 704 | |||
| GenBank mRNA | BC061858.1 | - | 6 | 699 | 5598495 | |
| CK365398.1 | - | 3 | 290 | |||
| X60789.1 | - | 6 | 667 | |||
| X60790.1 | - | 2 | 78 | |||
| X74565.1 | - | 6 | 685 | |||

Ptbp1
Genomic Map