siRNA Details
| Gene Symbol: Ptbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271057.1 | NP_001257986.1 | 4 | 245 | ||
| NM_022516.5 | NP_071961.2 | 4 | 245 | |||
| GenBank mRNA | BC061858.1 | - | 4 | 240 | 5598495 | |
| CB772933.1 | - | 4 | 255 | |||
| FN803753.1 | - | 4 | 215 | |||
| X60789.1 | - | 4 | 208 | |||
| X74565.1 | - | 4 | 226 | |||

Ptbp1
Genomic Map