siRNA Details
Gene Symbol: Slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017222.2 | NP_058918.1 | 1 | 161 | ||
XM_008771390.1 | XP_008769612.1 | 1 | 300 | |||
GenBank mRNA | U07183.2 | - | 1 | 161 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017222.2 | NP_058918.1 | 1 | 161 | ||
XM_008771390.1 | XP_008769612.1 | 1 | 300 | |||
GenBank mRNA | U07183.2 | - | 1 | 161 |