siRNA Details
| Gene Symbol: Nfya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012865.1 | NP_036997.1 | 6 | 628 | ||
| XM_006244409.2 | XP_006244471.1 | 6 | 635 | |||
| XM_006244410.1 | XP_006244472.1 | 6 | 631 | |||
| XM_006244411.2 | XP_006244473.1 | 6 | 625 | |||
| XM_006244412.2 | XP_006244474.1 | 6 | 622 | |||
| XM_006244413.1 | XP_006244475.1 | 5 | 543 | |||
| XM_008766839.1 | XP_008765061.1 | 5 | 531 | |||
| GenBank mRNA | FQ210330.1 | - | 5 | 548 | ||
| FQ223023.1 | - | 6 | 640 | |||
| M34238.1 | - | 6 | 628 | |||

Nfya
Genomic Map