siRNA Details
| Gene Symbol: Pctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017225.2 | NP_058921.1 | 2, 3 | 251 | ||
| XM_006247124.2 | XP_006247186.1 | 2, 3 | 237 | |||
| XM_006247125.2 | XP_006247187.1 | 2, 3 | 319 | |||
| XM_008767982.1 | XP_008766204.1 | 4, 5 | 458 | |||
| XM_008767983.1 | XP_008766205.1 | 4, 5 | 412 | |||
| GenBank mRNA | AF040261.1 | - | 2, 3 | 195 | ||
| BC078854.1 | - | 2, 3 | 256 | 7113616 | ||

Pctp
Genomic Map