siRNA Details
| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017227.1 | NP_058923.1 | 6 | 569 | ||
| XM_006239145.2 | XP_006239207.1 | 6 | 590 | |||
| GenBank mRNA | AB008803.1 | - | 6 | 569 | ||
| AB010999.1 | - | 6 | 569 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi4| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017227.1 | NP_058923.1 | 6 | 569 | ||
| XM_006239145.2 | XP_006239207.1 | 6 | 590 | |||
| GenBank mRNA | AB008803.1 | - | 6 | 569 | ||
| AB010999.1 | - | 6 | 569 | |||