siRNA Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017229.1 | NP_058925.1 | 6 | 1608 | ||
| GenBank mRNA | Z22867.1 | - | 6 | 1608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde3b| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017229.1 | NP_058925.1 | 6 | 1608 | ||
| GenBank mRNA | Z22867.1 | - | 6 | 1608 | ||