siRNA Details
| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193568.1 | NP_001180497.1 | 10, 11 | 1197 | ||
| NM_001193569.1 | NP_001180498.1 | 10, 11 | 1135 | |||
| NM_019232.3 | NP_062105.2 | 10, 11 | 1109 | |||
| XM_006227723.1 | XP_006227785.1 | 12, 13 | 1795 | |||
| XM_008758606.1 | XP_008756828.1 | 10, 11 | 1305 | |||
| GenBank mRNA | BC078843.1 | - | 10, 11 | 1063 | 7130800 | |
| L01624.1 | - | 10, 11 | 1111 | |||

Sgk1
Genomic Map